Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Chloropyrazolo[1,5-a]pyrimidine, 95%, Acros Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
58347-49-2
C6H4ClN3
153.57
MFCD04035684
YCZQHXPIKQHABJ-UHFFFAOYSA-N
7-chloropyrazolo 1,5-a pyrimidine,pyrazolo 1,5-a pyrimidine, 7-chloro,7-chloro-pyrazolo 1,5-a pyrimidine,zlchem 1087,7-chloropyrazolo 1?5-a pyrimidine,7-chloranylpyrazolo 1,5-a pyrimidine,7-chloro-8-hydropyrazolo 1,5-a pyrimidine
12281647
7-chloropyrazolo[1,5-a]pyrimidine
C1=C2N=CC=C(N2N=C1)Cl
58347-49-2
C6H4ClN3
153.57
MFCD04035684
YCZQHXPIKQHABJ-UHFFFAOYSA-N
7-chloropyrazolo 1,5-a pyrimidine,pyrazolo 1,5-a pyrimidine, 7-chloro,7-chloro-pyrazolo 1,5-a pyrimidine,zlchem 1087,7-chloropyrazolo 1?5-a pyrimidine,7-chloranylpyrazolo 1,5-a pyrimidine,7-chloro-8-hydropyrazolo 1,5-a pyrimidine
12281647
7-chloropyrazolo[1,5-a]pyrimidine
C1=C2N=CC=C(N2N=C1)Cl
Color Brown to White
Melting Point 98.0°C to 102.0°C
Quantity 1g
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Formula Weight 153.57
Physical Form Powder
Percent Purity 95%
Chemical Name or Material 7-Chloropyrazolo[1,5-a]pyrimidine
Assay 95%
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off im

GHS Signal Word: Danger