Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroquine Phosphate, EP, 98.5-101%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
500 g
2.5 kg
Packaging:
Amber Glass Bottle
Poly Pail
50-63-5
C18H32ClN3O8P2
515.86
QKICWELGRMTQCR-UHFFFAOYNA-N
7-chloro-N-[5-(diethylamino)pentan-2-yl]quinolin-4-amine; bis(phosphoric acid)
OP(O)(O)=O.OP(O)(O)=O.CCN(CC)CCCC(C)NC1=C2C=CC(Cl)=CC2=NC=C1
50-63-5
C18H32ClN3O8P2
515.86
QKICWELGRMTQCR-UHFFFAOYNA-N
7-chloro-N-[5-(diethylamino)pentan-2-yl]quinolin-4-amine; bis(phosphoric acid)
OP(O)(O)=O.OP(O)(O)=O.CCN(CC)CCCC(C)NC1=C2C=CC(Cl)=CC2=NC=C1

C1960, 50-63-5, C18H26ClN3•2H3PO4

Spectrum™ Chloroquine Phosphate, EP is used as a bacteriacide, an algaecide and as an antimalarial. All Spectrum Chemical EP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Percent Purity 98.5-101%
Loss on Drying 2%
Packaging Amber Glass Bottle
Quantity 5 g