Chlorosulfonic acid, typically 99%, Alfa Aesar™

Manufacturer: Alfa Aesar 87977D5

Quantity 3 x 1kg

 View more versions of this product

Catalog No. AA87977D5

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7790-94-5
    Physical Form Liquid
    Assay Percent Range typically 99%
    Molecular Formula HSO3Cl
    Formula Weight 116.52
    MDL Number MFCD00011523
    Chemical Name or Material Chlorosulfonic acid

    Specifications

    Formula Weight 116.52
    Quantity 3 x 1kg
    Color Yellow to brown
    UN Number 1754
    DOT Information DOT Class: 8, Packaging Group: I
    EINECS Number 232-234-6
    Physical Form Liquid
    Molecular Formula HSO3Cl
    CAS 7790-94-5
    Health Hazard 1 H314-H335 : Causes severe skin burns and eye damage. May cause respiratory irritation.
    Appearance Yellow to brown liquid
    Odor Odorless
    Assay Percent Range typically 99%
    MDL Number MFCD00011523
    Chemical Name or Material Chlorosulfonic acid

    H314-H335 : Causes severe skin burns and eye damage. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit