Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorosulfonylacetyl chloride, 95%, Acros Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
4025-77-8
C2H2Cl2O3S
176.995
MFCD00007457
MCFSNYMQISXQTF-UHFFFAOYSA-N
chlorosulfonylacetyl chloride,acetyl chloride, chlorosulfonyl,acetyl chloride, 2-chlorosulfonyl,2-chlorosulfonyl acetyl chloride,chlorosulfonyl acetyl chloride,chlorosulphonyl acetyl chloride,chlorosulfonylacetylchloride,acmc-20algk,c2h2cl2so3
77642
2-chlorosulfonylacetyl chloride
C(C(=O)Cl)S(=O)(=O)Cl
4025-77-8
C2H2Cl2O3S
176.995
MFCD00007457
MCFSNYMQISXQTF-UHFFFAOYSA-N
chlorosulfonylacetyl chloride,acetyl chloride, chlorosulfonyl,acetyl chloride, 2-chlorosulfonyl,2-chlorosulfonyl acetyl chloride,chlorosulfonyl acetyl chloride,chlorosulphonyl acetyl chloride,chlorosulfonylacetylchloride,acmc-20algk,c2h2cl2so3
77642
2-chlorosulfonylacetyl chloride
C(C(=O)Cl)S(=O)(=O)Cl
Alkylation reagent
Color White to Brown
Boiling Point 71.0°C to 72.0°C (1.0mmHg)
Quantity 1g
Packaging Glass bottle
Infrared Spectrum Authentic
Flash Point >110°C
Density 1.6690g/mL
Assay Percent Range 95%
Linear Formula ClSO2CH2COCl
UN Number 3265
Solubility Information Solubility in water: reacts.
Formula Weight 177.01
Physical Form Powder, Crystals and/or Chunks
Percent Purity 95%
Chemical Name or Material chlorosulfonylacetyl chloride
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Reacts violently with water.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger