Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Chlorotri-n-propylsilane, 98%

Manufacturer:  Alfa Aesar™ B2206822

Quantity 100g

 View more versions of this product

Catalog No. AAB2206822


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

995-25-5
192.81
UN2987
Tri-n-propylchlorosilane; Tri-n-propylsilyl chloride
(CH3CH2CH2)3SiCl
MFCD00000508
1737489
Chlorotri-n-propylsilane

Specifications

995-25-5
192.81
UN2987
Tri-n-propylchlorosilane; Tri-n-propylsilyl chloride
199° to 201°C
70°C (158°F)
98%
(CH3CH2CH2)3SiCl
MFCD00000508
1737489
Acrid
100g
Chlorotri-n-propylsilane

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit