Alfa Aesar™ Chlorotri-n-propylsilane, 98%

Manufacturer: Alfa Aesar™ B2206814

Quantity 25g

 View more versions of this product

Catalog No. AAB2206814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 995-25-5
    Molecular Formula (CH3CH2CH2)3SiCl
    Formula Weight 192.81
    MDL Number MFCD00000508
    UN Number UN2987
    Beilstein 1737489
    Synonym Tri-n-propylchlorosilane; Tri-n-propylsilyl chloride
    Chemical Name or Material Chlorotri-n-propylsilane

    Specifications

    Formula Weight 192.81
    TSCA TSCA
    Odor Acrid
    Molecular Formula (CH3CH2CH2)3SiCl
    MDL Number MFCD00000508
    CAS 995-25-5
    UN Number UN2987
    Beilstein 1737489
    Synonym Tri-n-propylchlorosilane; Tri-n-propylsilyl chloride
    Boiling Point 199° to 201°C
    Quantity 25g
    Flash Point 70°C (158°F)
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.
    Chemical Name or Material Chlorotri-n-propylsilane
    Assay 98%

    GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit