Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Chlorotri-n-propylsilane, 98%

Click to view available options
Quantity:
5g
25g
995-25-5
C9H21ClSi
192.802
MFCD00000508
ACTAPAGNZPZLEF-UHFFFAOYSA-N
tri-n-propylchlorosilane,tripropylchlorosilane,silane, chlorotripropyl,chlorotri-n-propylsilane,chloro tripropyl silane,chloro-tri-n-propylsilane,chlorotripropylsilane,chloro tri-n-propyl silane
70445
chloro(tripropyl)silane
CCC[Si](CCC)(CCC)Cl
995-25-5
C9H21ClSi
192.802
MFCD00000508
ACTAPAGNZPZLEF-UHFFFAOYSA-N
tri-n-propylchlorosilane,tripropylchlorosilane,silane, chlorotripropyl,chlorotri-n-propylsilane,chloro tripropyl silane,chloro-tri-n-propylsilane,chlorotripropylsilane,chloro tri-n-propyl silane
70445
chloro(tripropyl)silane
CCC[Si](CCC)(CCC)Cl
UN Number UN2987
Beilstein 1737489
Formula Weight 192.81
Percent Purity 98%
Odor Acrid
Boiling Point 199°C to 201°C
Quantity 5g
Flash Point 70°C (158°F)
Chemical Name or Material Chlorotri-n-propylsilane

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.

TSCA : TSCA