Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorotriethylsilane, 99%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
100mL
500mL
Packaging:
Glass bottle
994-30-9
C6H15ClSi
150.72
DCFKHNIGBAHNSS-UHFFFAOYSA-N
triethylchlorosilane,silane, chlorotriethyl,triethylsilyl chloride,chloro triethyl silane,unii-q1a687m1py,triethyl silyl chloride,triethychlorosilane,chlorotrietylsilane,clsiet3,et3sicl
13819
chloro(triethyl)silane
CC[Si](CC)(CC)Cl
994-30-9
C6H15ClSi
150.72
DCFKHNIGBAHNSS-UHFFFAOYSA-N
triethylchlorosilane,silane, chlorotriethyl,triethylsilyl chloride,chloro triethyl silane,unii-q1a687m1py,triethyl silyl chloride,triethychlorosilane,chlorotrietylsilane,clsiet3,et3sicl
13819
chloro(triethyl)silane
CC[Si](CC)(CC)Cl
Silylation reagent
Specific Gravity 0.89
Boiling Point 142.0°C to 144.0°C
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 100mL
Refractive Index 1.4300 to 1.4320
Flash Point 29°C
Density 0.8900g/mL
Color Colorless to Yellow
Assay Percent Range 98.5% min. (GC)
Linear Formula (C2H5)3SiCl
Beilstein 04,627
Fieser 13,165
Merck Index 15,2179
Solubility Information Solubility in water: decomposes
Formula Weight 150.72
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Chlorotriethylsilane
Show More Show Less

GHS Signal Word: Danger

EINECSNumber : 213-615-6