Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorotriisopropylsilane, 95%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
13154-24-0
C9H21ClSi
192.81
MFCD00009656
KQIADDMXRMTWHZ-UHFFFAOYSA-N
triisopropylsilyl chloride,chlorotriisopropylsilane,triisopropylchlorosilane,chlorotris propan-2-yl silane,chloro triisopropyl silane,silane, chlorotris 1-methylethyl,tipscl,triisopropylsilylchloride,triisopropyl chlorosilane,triisopropylchlorosilane, redistilled
139400
chloro-tri(propan-2-yl)silane
CC(C)[Si](C(C)C)(C(C)C)Cl
13154-24-0
C9H21ClSi
192.81
MFCD00009656
KQIADDMXRMTWHZ-UHFFFAOYSA-N
triisopropylsilyl chloride,chlorotriisopropylsilane,triisopropylchlorosilane,chlorotris propan-2-yl silane,chloro triisopropyl silane,silane, chlorotris 1-methylethyl,tipscl,triisopropylsilylchloride,triisopropyl chlorosilane,triisopropylchlorosilane, redistilled
139400
chloro-tri(propan-2-yl)silane
CC(C)[Si](C(C)C)(C(C)C)Cl
Refractive Index 1.4520 to 1.454
Flash Point 63°C
Boiling Point 198°C
Specific Gravity 0.902
Quantity 5g
Packaging Glass bottle
Color Colorless to Yellow
Density 0.9020g/mL
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Linear Formula [(CH3)2CH]3SiCl
Solubility Information Solubility in water: reacts
Formula Weight 192.81
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material Chlorotriisopropylsilane
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger