Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorotriphenylsilane 95%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
76-86-8
C18H15ClSi
294.85
MFCD00000496
MNKYQPOFRKPUAE-UHFFFAOYSA-N
triphenylsilyl chloride,triphenylchlorosilane,silane, chlorotriphenyl,triphenylsilicon chloride,chloro triphenyl silane,triphenylsilylchloride,chloro-tri phenyl silane,benzene, 1,1',1-chlorosilylidyne tris,clsiph3,ph3sicl
6458
chloro(triphenyl)silane
C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(C3=CC=CC=C3)Cl
76-86-8
C18H15ClSi
294.85
MFCD00000496
MNKYQPOFRKPUAE-UHFFFAOYSA-N
triphenylsilyl chloride,triphenylchlorosilane,silane, chlorotriphenyl,triphenylsilicon chloride,chloro triphenyl silane,triphenylsilylchloride,chloro-tri phenyl silane,benzene, 1,1',1-chlorosilylidyne tris,clsiph3,ph3sicl
6458
chloro(triphenyl)silane
C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(C3=CC=CC=C3)Cl
Boiling Point 378°C
Melting Point 91°C to 96°C
Quantity 100g
Packaging Glass bottle
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 95%
Linear Formula (C6H5)3SiCl
Beilstein 16, 905
Fieser 12,279
Solubility Information Solubility in water: decomposes
Formula Weight 294.85
Physical Form Crystalline Powder and Crystals
Percent Purity ≥94%
Chemical Name or Material Chlorotriphenylsilane, 95%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 200-989-0

RTECSNumber : VV2720000

TSCA : TSCA