Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

chlorotripyrrolidinophosphonium hexafluorophosphate, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
133894-48-1
C12H24ClN3P·F6P
421.73
BSCYRXJVGSZNKX-UHFFFAOYSA-N
pyclop,chlorotripyrrolidinophosphonium hexafluorophosphate,chlorotripyrrolidinophosphonium hexa-fluorophosphate,chlorotri 1-pyrrolidinyl phosphonium hexafluorophosphate, 98+%,pyciop,ref dupl: pyclop,chlorotripyrrolidino,acmc-1c01w,ksc177a1n,chlorotripyrrolidinophosphoniumhexafluorophosphat
16211753
chloro(tripyrrolidin-1-yl)phosphanium;hexafluorophosphate
C1CCN(C1)[P+](N2CCCC2)(N3CCCC3)Cl.F[P-](F)(F)(F)(F)F
133894-48-1
C12H24ClN3P·F6P
421.73
BSCYRXJVGSZNKX-UHFFFAOYSA-N
pyclop,chlorotripyrrolidinophosphonium hexafluorophosphate,chlorotripyrrolidinophosphonium hexa-fluorophosphate,chlorotri 1-pyrrolidinyl phosphonium hexafluorophosphate, 98+%,pyciop,ref dupl: pyclop,chlorotripyrrolidino,acmc-1c01w,ksc177a1n,chlorotripyrrolidinophosphoniumhexafluorophosphat
16211753
chloro(tripyrrolidin-1-yl)phosphanium;hexafluorophosphate
C1CCN(C1)[P+](N2CCCC2)(N3CCCC3)Cl.F[P-](F)(F)(F)(F)F
Coupling reagent
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 421.73
Physical Form Crystalline Powder
Percent Purity 98%
Elemental Analysis 33.33 to 35.03% C, 9.71 to 10.21% N
Infrared Spectrum Authentic
Packaging Glass bottle
Color White to Yellow
Melting Point 145.0°C to 148.0°C
Quantity 25g
Chemical Name or Material Chlorotripyrrolidinophosphonium hexafluorophosphate
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger