Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chlorotripyrrolidinophosphonium Hexafluorophosphate 98.0+%, TCI America™
Click to view available options
Quantity:
5 g
25 g
Specifications
Melting Point | 138°C |
Color | White-Yellow |
UN Number | 1759 |
Quantity | 25 g |
Formula Weight | 421.73 |
Percent Purity | ≥98.0% (HPLC,T) |
Physical Form | Crystalline Powder |
Chemical Name or Material | Chlorotripyrrolidinophosphonium Hexafluorophosphate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator