Alfa Aesar™ Chlorotris(triphenylphosphine)cobalt(I), 97%

Manufacturer: Alfa Aesar™ H5539514

Quantity 25g

 View more versions of this product

Catalog No. AAH5539514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 26305-75-9
    Molecular Formula C54H45ClCoP3
    Formula Weight 881.26
    MDL Number MFCD00015864
    Chemical Name or Material Chlorotris(triphenylphosphine)cobalt(I)

    Specifications

    Formula Weight 881.26
    Molecular Formula C54H45ClCoP3
    MDL Number MFCD00015864
    CAS 26305-75-9
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H317
    Toxic if swallowed.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P363-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 135° to 139°C
    Quantity 25g
    Chemical Name or Material Chlorotris(triphenylphosphine)cobalt(I)
    Assay 97%

    Description

    Applications
    A stoichiometric reducing agent that is used in the radical dimerization of halogenated organic molecules.

    Solubility
    Insoluble in water.

    Notes
    Air & Moisture Sensitive.

    GHS H Statement
    H301-H317
    Toxic if swallowed.
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P301+P310-P363-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit