Alfa Aesar™ Chlorotris(triphenylphosphine)rhodium(I), 97%

Manufacturer: Alfa Aesar™ 1046803

Quantity 1g

 View more versions of this product

Catalog No. AA1046803

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 14694-95-2
    Linear Formula RhCl[(C6H5)3P]3
    Formula Weight 925.23
    MDL Number MFCD00010016
    Merck Index 14,10047
    Synonym Wilkinson's catalyst; Tris(triphenylphosphine)rhodium(I) chloride
    Chemical Name or Material Chlorotris(triphenylphosphine)rhodium(I)

    Specifications

    Formula Weight 925.23
    Solubility Information Soluble in most solvents (e.g. benzene, ethanol, chloroform, dichloromethane) but with phosphine dissociation; Reacts with O2 in solution
    Merck Index 14,10047
    TSCA TSCA
    Odor Odorless
    Physical Form Micro Crystals
    MDL Number MFCD00010016
    Synonym Wilkinson's catalyst; Tris(triphenylphosphine)rhodium(I) chloride
    Storage Note 1 2°C to 8°C
    Color Dark red
    Melting Point ≈250°C (dec.)
    Quantity 1g
    Linear Formula RhCl[(C6H5)3P]3
    CAS 14694-95-2
    Chemical Name or Material Chlorotris(triphenylphosphine)rhodium(I)
    Assay 97%

    2°C to 8°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit