Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorpromazine Hydrochloride, USP, 98-101.5%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
1 kg
12 kg
Packaging:
Amber Glass Bottle
Fiber Drum
69-09-0
C17H20Cl2N2S
355.32
FBSMERQALIEGJT-UHFFFAOYSA-N
hydrogen [3-(2-chloro-10H-phenothiazin-10-yl)propyl]dimethylamine chloride
[H+].[Cl-].CN(C)CCCN1C2=CC=CC=C2SC2=CC=C(Cl)C=C12
69-09-0
C17H20Cl2N2S
355.32
FBSMERQALIEGJT-UHFFFAOYSA-N
hydrogen [3-(2-chloro-10H-phenothiazin-10-yl)propyl]dimethylamine chloride
[H+].[Cl-].CN(C)CCCN1C2=CC=CC=C2SC2=CC=C(Cl)C=C12

CH132, 69-09-0, C17H19ClN2S•HCl

Spectrum™ Chlorpromazine Hydrochloride, USP is a dopamine antagonist used to treat schizophrenia and has served as the prototype for the phenothiazine class of drugs. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Loss on Drying 0.5%
Ignition Residue 0.1%
Melting Range 195-198°C
Quantity 25 g
Packaging Amber Glass Bottle
CAS Max % 100%
UN Number UN2811
Percent Purity 98-101.5%