Learn More
Chlorpyrifos Rabbit anti-Nonspecies specific, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP288859
Catalog No. NB036478
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Chlorpyrifos Polyclonal antibody specifically detects Chlorpyrifos in Nonspecies specific samples. It is validated for ELISA, Affinity Purification
Specifications
Specifications
Chlorpyrifos
Unconjugated
Whole antiserum
chlorpyrifos-ethyl, Clorpyrifos
BSA-Chlorpyrifos Conjugate
Whole Antiserum
RUO
Primary
Nonspecies specific
ELISA, Affinity Purification
ELISA 1:2000, Immunoaffinity Purification
Antiserum
Rabbit
IgG
0.1 mL
Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles.
Polyclonal