Learn More
CTL Scientific Supply Corp CHLORTESMO,200 STRIPS
SDP
Supplier:  CTL Scientific Supply Corp 90603
Encompass_Preferred
CHLORTESMO is a test paper for the determination of free halogens (chlorine, bromine, iodine). Free nitrous acid HNO 2 (not nitrite ions) interferes, but can be destroyed by additions of amidosulphuric acid. Box of 200 strips (each strip 20 x 70 mm). Limit of sensitivity: 1 mg/l chlorine
Catalog No. 50-117-3279