Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorthalidone, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
77-36-1
C14H11ClN2O4S
338.76
JIVPVXMEBJLZRO-UHFFFAOYNA-N
2-chloro-5-(1-hydroxy-3-oxo-2,3-dihydro-1H-isoindol-1-yl)benzene-1-sulfonamide
NS(=O)(=O)C1=CC(=CC=C1Cl)C1(O)NC(=O)C2=CC=CC=C12
77-36-1
C14H11ClN2O4S
338.76
JIVPVXMEBJLZRO-UHFFFAOYNA-N
2-chloro-5-(1-hydroxy-3-oxo-2,3-dihydro-1H-isoindol-1-yl)benzene-1-sulfonamide
NS(=O)(=O)C1=CC(=CC=C1Cl)C1(O)NC(=O)C2=CC=CC=C12

C4010, 77-36-1, C14H11CIN2O4S

Spectrum™ Chlorthalidone, USP
Packaging Poly Pail
Ignition Residue 0.001
Loss on Drying 0.004
Quantity 5 kg
CAS Max % 1
Percent Purity 98 to 102%