Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHMP2A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5119777

This item is not returnable.
View return policy
Description
Positive Control: SW480 cell lysate, A431 cell lysate, HepG2 cell lysate, rat bladder tissue. Subcellular Location: Late endosome membrane.
CHMP2A Polyclonal antibody specifically detects CHMP2A in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
CHMP2A | |
Polyclonal | |
Unconjugated | |
CHMP2A | |
1500016L11Rik; BC2; BC-2; Charged multivesicular body protein 2a; CHMP2; CHMP2A; chromatin modifying protein 2A; chromatin-modifying protein 2a; fj10d05; hypothetical protein LOC523847; mVps2; putative breast adenocarcinoma marker; putative breast adenocarcinoma marker (32kD); putative breast adenocarcinoma marker BC-2; RGD1305050; Vacuolar protein sorting-associated protein 2; vacuolar protein sorting-associated protein 2-1; vacuolar protein-sorting-associated protein 2; VPS2; VPS2 homolog A; vps2-1; VPS2A; wu:fi18h03; wu:fj10d05; zgc:55361 | |
Synthetic peptide within N-terminal human CHMP2A | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
O43633, Q9DB34 | |
Rabbit | |
Affinity chromatography | |
RUO | |
27243, 365191, 68953 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title