Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHMP4B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP191782
Description
CHMP4B Polyclonal specifically detects CHMP4B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CHMP4B | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
C20orf178, charged multivesicular body protein 4b, CHMP4A, CHMP4b, chromatin modifying protein 4B, Chromatin-modifying protein 4b, chromosome 20 open reading frame 178, dJ553F4.4, hSnf7-2, hVps32-2, SHAX1, SNF7, SNF7 homolog associated with Alix 1, Snf7 homologue associated with Alix 1, SNF7-2CTPP3, Vacuolar protein sorting-associated protein 32-2, vacuolar protein-sorting-associated protein 32-2, Vps32-2, VPS32B | |
Rabbit | |
Affinity Purified | |
RUO | |
128866 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
CHMP4B | |
This antibody was developed against Recombinant Protein corresponding to amino acids:SVFGKLFGAGGGKAGKGGPTPQEAIQRLRDTEEMLSKK | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction