Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CHMP4B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562471
Description
Immunogen sequence: EQEELDKNLL EISGPETVPL PNVPSIALPS KPAKKKEEED DDMKELENWA GSV Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
CHMP4B is a member of the chromatin-modifying protein/charged multivesicular body protein (CHMP) protein family. The protein is part of the endosomal sorting complex required for transport (ESCRT) complex III (ESCRT-III), which functions in the sorting of endocytosed cell-surface receptors into multivesicular endosomes. The ESCRT machinery also functions in the final abscisson stage of cytokinesis and in the budding of enveloped viruses such as HIV-1. The three proteins of the CHMP4 subfamily interact with programmed cell death 6 interacting protein (PDCD6IP, also known as ALIX), which also functions in the ESCRT pathway. The CHMP4 proteins assemble into membrane-attached 5-nm filaments that form circular scaffolds and promote or stabilize outward budding. These polymers are proposed to help generate the luminal vesicles of multivesicular bodies.
Specifications
CHMP4B | |
Polyclonal | |
Unconjugated | |
CHMP4B | |
2010012F05Rik; C20orf178; C76846; charged multivesicular body protein 4b; CHMP4A; CHMP4B; chromatin modifying protein 4B; chromatin-modifying protein 4b; CTPP3; CTRCT31; dJ553F4.4; hSnf7-2; hVps32-2; RGD1309846; RGD1565889; Shax1; SNF7; SNF7 homolog associated with Alix 1; Snf7 homologue associated with Alix 1; Snf7-2; Vacuolar protein sorting-associated protein 32-2; vacuolar protein-sorting-associated protein 32-2; Vps32-2; VPS32B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
128866 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H444 | |
CHMP4B | |
Recombinant protein corresponding to Human CHMP4B. Recombinant protein control fragment (Product #RP-104187). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction