Learn More
CHMP7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 164241AP
Catalog No. 50-173-0936
Add to cart
Description
Description
CHMP7 gene ontology annotations related to this gene include vacuolar transport.
Specifications
Specifications
CHMP7
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8R1T1, Q8WUX9
CHMP family, member 7, CHMP7, Chromatin modifying protein 7
Rabbit
IgG
150 μL
-20°C
Primary
105513, 364419, 91782
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CHMP7
Liquid
Chmp7
CHMP7 Fusion Protein Ag9625
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat