Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHN 1 Mouse anti-Human, DyLight™ 405, Clone: OTI2G6, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP270411V
Description
CHN 1 Monoclonal antibody specifically detects CHN 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.Specifications
CHN 1 | |
OTI2G6 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
50mM Sodium Borate with 0.05% Sodium Azide | |
1123.0 | |
Mouse | |
IgG2a | |
Primary | |
0.1 mL | |
Human |
Western Blot,Immunohistochemistry,Immunocytochemistry,Immunofluorescence | |
DyLight 405 | |
Affinity Purified,Conjugated | |
a2-chimaerin, A-chimaerin, alpha-chimerin, ARHGAP2RhoGAP2, chimerin (chimaerin) 1, CHNDuane retraction syndrome 2, DURS2, NC, N-chimaerin, N-chimerin, Rho GTPase-activating protein 2, RHOGAP2 | |
CHN1 | |
Full length recombinant protein of human CHN 1 (NP_001813) produced in HEK293T cells. | |
Monoclonal | |
Immunogen affinity purified | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title