Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHN1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549289

Catalog No. PA549289

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with mouse based on sequence homology.

GTPase-activating protein for p21-rac and a phorbol ester receptor. Involved in the assembly of neuronal locomotor circuits as a direct effector of EPHA4 in axon guidance.
Specifications

Specifications

CHN1
Polyclonal
PBS with 0.09% sodium azide
P15882
N-chimaerin, A-chimaerin, Alpha-chimerin, N-chimerin, NC, Rho GTPase-activating protein 2, CHN1, ARHGAP2, CHN
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1123
Western Blot
Unconjugated
CHN1
Liquid
CHN1
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human CHN1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit