Learn More
Gibco™ CHO Cell Culture Medium
A complete animal origin-free (AOF) and serum-free, ready-to-use formulation containing a plant hydrolysate for maximum productivity.
Manufacturer:  Gibco™ 220253
Catalog No. 22-025-3
Add to cart
Description
Description
  • This medium is formulated to support cell growth and production of antibodies and recombinant proteins in suspension CHO cell cultures
  • Can be used in various culture vessels from T-flasks, shaker flasks, and roller bottles to spinners and bioreactors
  • The medium can be used in research or manufacturing applications
  • It has been developed after careful screening of a wide variety of hydrolysates for protein yield increases in CHO cells
  • Appropriate for suspension CHO cells
  • It is also appropriate for a selection system such as glutamine synthetase (GS), which does not require the selective pressure of hypoxanthine/thymidine (HT)-deficient medium
Specifications
Specifications
Animal origin-free
Cell culture media
Liquid
1 L
Mammalian
Supports cell growth and production of antibodies and recombinant proteins in suspension CHO cell cultures
Protect from light, 2°C to 8°C
No glutamine
Liquid