Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cholecalciferol, Crystal, USP, 97-103%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
1 kg
Packaging:
Amber Glass Bottle
67-97-0
C27H44O
384.65
QYSXJUFSXHHAJI-FVUVGDFOSA-N
(1S,3E)-3-{2-[(1R,3aS,4E,7aR)-7a-methyl-1-[(2R)-6-methylheptan-2-yl]-octahydro-1H-inden-4-ylidene]ethylidene}-4-methylidenecyclohexan-1-ol
CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C1/C[C@@H](O)CCC1=C
67-97-0
C27H44O
384.65
QYSXJUFSXHHAJI-FVUVGDFOSA-N
(1S,3E)-3-{2-[(1R,3aS,4E,7aR)-7a-methyl-1-[(2R)-6-methylheptan-2-yl]-octahydro-1H-inden-4-ylidene]ethylidene}-4-methylidenecyclohexan-1-ol
CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C1/C[C@@H](O)CCC1=C

CH134, 67-97-0, C27H44O

Spectrum™ Cholecalciferol, Crystal, USP is also known as toxiferol and is a form of vitamin D. It is used in vitamin supplements. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Specific Rotation +105° to +112°
Quantity 25 g
CAS Max % 1
UN Number UN2811
Percent Purity 97 to 103%