Learn More
Alfa Aesar™ Cholecystokinin (26-33), Non-sulfated
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66378MCR
 View more versions of this product
Catalog No. AAJ66378MCR
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
1063.20
1mg
Asp-Tyr-Met-Gly-Trp-Met-Asp-Phe-NH2
-30° to -10°C
Cholecystokinin (26-33), non-sulfated
HPLC
Solid
RUO
CCK Octapeptide (26-33), non-sulfated