Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alere Cassette Cartridges for Cholestech LDX™ System Available on GSA/VA Contract for Federal Government customers only.

Cassette cartridges for use with Cholestech LDX cholesterol analyzer.

Manufacturer:  Alere 10989

 View more versions of this product

Catalog No. 23-762-960

 

Due to product restrictions, please to purchase or view availability for this product.


Specifications

Description

  • Rapid: Yields results in 5 minutes
  • Flexible: fingerstick or venous whole blood
  • Convenient: Sample size is 40μL
  • CLIA-waived (for whole blood)
Description & Specifications

Specifications

Test Cassette
Blood
CLIA waived/professional use only
10 Individually wrapped test cassettes and package insert
36° to 46°F
40μL
Cholestech LDX™ Cholesterol analyzer
Lipid Profile
5 min.
2.22° to 7.77°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit