Alere MiniPet™ Pipette for Cholesterol Analyzers

For use with Cholestech LDX lipid monitoring system.

Manufacturer: Alere 11846

 View more versions of this product

Catalog No. 23-763-003

  Check Availability
Add to Cart

    Specifications

    For Use With (Equipment) Cholestech LDX™ lipid monitoring system
    Product Type MiniPet™ pipette
    Sample Type Venous Whole Blood
    Capacity (Metric) 35μL

    For use with Cholestech LDX lipid monitoring system.

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit