Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cholesterol, Affymetrix/USB™

Click to view available options
Quantity:
5g
100g
Packaging:
Plastic bottle
57-88-5
C27H46O
386.664
MFCD00003646
HVYWMOMLDIMFJA-DPAQBDIFSA-N
cholesterol,cholesterin,cholest-5-en-3beta-ol,cholesteryl alcohol,cholestrin,cordulan,dusoline,dusoran,provitamin d,cholesterine
5997
CHEBI:16113
(3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
CC(C)CCCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)O)C)C
57-88-5
C27H46O
386.664
MFCD00003646
HVYWMOMLDIMFJA-DPAQBDIFSA-N
cholesterol,cholesterin,cholest-5-en-3beta-ol,cholesteryl alcohol,cholestrin,cordulan,dusoline,dusoran,provitamin d,cholesterine
5997
CHEBI:16113
(3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
CC(C)CCCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)O)C)C
Packaging Plastic bottle
Specific Gravity 1.067
Melting Point 147°C to 150°C
Quantity 5g
Boiling Point 360°C
Formula Weight 386.65
Physical Form Powder
Chemical Name or Material Cholesterol