Learn More
Thermo Scientific Chemicals Cholesterol Esterase, from porcine pancreas
Supplier: Thermo Scientific Chemicals J64282EXU
Description
Cholesterol Esterase is used for enzymatic determination of total cholesterol when coupled with cholesterol oxidize in clinical analysis. Cholesterol esterase (CE) is a reversible enzyme that can hydrolyze or synthesize fatty acid esters of cholesterol and other sterols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
500 U | |
440 kd | |
Lyophilized |
Safety and Handling
EINECSNumber : 232-808-6
Recommended Storage : Store at -20°C