Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cholesterol, Fisher Chemical

Click to view available options
Quantity:
500g
57-88-5
C27H46O
386.664
HVYWMOMLDIMFJA-DPAQBDIFSA-N
cholesterol, cholesterin, cholest-5-en-3beta-ol, cholesteryl alcohol, cholestrin, cordulan, dusoline, dusoran, provitamin d, cholesterine
5997
CHEBI:16113
(3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
CC(C)CCCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)O)C)C
57-88-5
C27H46O
386.664
HVYWMOMLDIMFJA-DPAQBDIFSA-N
cholesterol, cholesterin, cholest-5-en-3beta-ol, cholesteryl alcohol, cholestrin, cordulan, dusoline, dusoran, provitamin d, cholesterine
5997
CHEBI:16113
(3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
CC(C)CCCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)O)C)C
Odor Odorless
Color White
Density 1.067
Melting Point 148.9°C
Quantity 500g
UN Number 1845
Formula Weight 385.31
Physical Form Solid
Chemical Name or Material Cholesterol (USP)

DOTInformation : Hazard Class 9, Packing group III

Recommended Storage : Keep containers tightly closed in a dry, cool and well-ventilated place