Alfa Aesar™ Cholesteryl chloride, 98%

Manufacturer: Alfa Aesar™ A1043336

Quantity 500g

 View more versions of this product

Catalog No. AAA1043336

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 910-31-6
    Molecular Formula C27H45Cl
    Formula Weight 405.11
    MDL Number MFCD00003632
    Beilstein 2703655
    Chemical Name or Material Cholesteryl chloride

    Specifications

    Formula Weight 405.11
    TSCA TSCA
    Molecular Formula C27H45Cl
    CAS 910-31-6
    Beilstein 2703655
    Optical Rotation −26° (c=1 in benzene)
    MDL Number MFCD00003632
    Melting Point 94° to 96°C
    Quantity 500g
    Chemical Name or Material Cholesteryl chloride
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit