Alfa Aesar™ Cholesteryl oleate

Manufacturer: Alfa Aesar™ A1137806

Quantity 5g

 View more versions of this product

Catalog No. AAA1137806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 303-43-5
    Molecular Formula C45H78O2
    Formula Weight 651.12
    MDL Number MFCD00003645
    Beilstein 2343071
    Synonym Oleic acid cholesteryl ester
    Chemical Name or Material Cholesteryl oleate

    Specifications

    Formula Weight 651.12
    CAS 303-43-5
    Synonym Oleic acid cholesteryl ester
    Molecular Formula C45H78O2
    MDL Number MFCD00003645
    Quantity 5g
    Odor Odorless
    TSCA TSCA
    Beilstein 2343071
    Melting Point 44° to 47°C
    Optical Rotation −24° (c=1 in chloroform)
    Chemical Name or Material Cholesteryl oleate

    Description

    Applications
    Cholesteryl oleate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff. It is also used in chemical research.

    Solubility
    It is soluble in chloroform: 0.1 g/Ml.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Incompatible to strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit