Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cholestyramine Resin, USP, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
500 g
5 kg
50 kg
Packaging:
Amber Glass Bottle
Drum
Poly Pail
11041-12-6
C21H30ClN
331.93
KNDHRUPPBXRELB-UHFFFAOYNA-M
4-[3-(4-ethylphenyl)butyl]-N,N,N-trimethylanilinium chloride
[Cl-].CCC1=CC=C(C=C1)C(C)CCC1=CC=C(C=C1)[N+](C)(C)C
11041-12-6
C21H30ClN
331.93
KNDHRUPPBXRELB-UHFFFAOYNA-M
4-[3-(4-ethylphenyl)butyl]-N,N,N-trimethylanilinium chloride
[Cl-].CCC1=CC=C(C=C1)C(C)CCC1=CC=C(C=C1)[N+](C)(C)C

C1734, 11041-12-6

Spectrum™ Cholestyramine Resin, USP is a cholesterol lowering agent. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Quantity 5 g
Loss on Drying 12%
Ignition Residue 0.1%
Packaging Amber Glass Bottle
CAS Max % 100%