Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cholic acid, 98+%

Click to view available options
Quantity:
25g
100g
500g
81-25-4
C24H40O5
408.579
MFCD00003672
BHQCQFFYRZLCQQ-OELDTZBJSA-N
cholic acid,cholalic acid,cholate,cholalin,colalin,cholsaeure,cholbam,cholicacid,cholic acid, 5beta,3alpha,7alpha,12alpha-trihydroxy-5beta-cholanic acid
221493
CHEBI:16359
(4R)-4-[(3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
CC(CCC(=O)O)C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C
81-25-4
C24H40O5
408.579
MFCD00003672
BHQCQFFYRZLCQQ-OELDTZBJSA-N
cholic acid,cholalic acid,cholate,cholalin,colalin,cholsaeure,cholbam,cholicacid,cholic acid, 5beta,3alpha,7alpha,12alpha-trihydroxy-5beta-cholanic acid
221493
CHEBI:16359
(4R)-4-[(3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
CC(CCC(=O)O)C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C
Applications
Cholic acid is used as a preservative for food. It is widely used as an intermediate in the manufacture of pharmaceuticals. It finds use in biological research. It helps in detergents to soften biological fats. It helps in preventing and melting gallstones.

Solubility
Soluble in alcohols, acetone, dimethylsulfoxide, methanol. Insoluble in water.

Notes
Stable. Incompatible with other materials such as strong oxidizing agents.
Optical Rotation +35 ± 2° (c=0.6 in ethanol)
Melting Point 199°C to 202°C
Quantity 25g
Beilstein 2822009
Merck Index 14,2203
Solubility Information Soluble in alcohols,acetone,dimethylsulfoxide,methanol. Insoluble in water.
Formula Weight 408.58
Percent Purity ≥98%
Chemical Name or Material Cholic acid

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 201-337-8

RTECSNumber : FZ9350000

TSCA : Yes

Recommended Storage : Ambient temperatures