Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cholic Acid Sodium Salt, ≥98.5%, MP Biomedicals™

Catalog Number p-4404522
Click to view available options
Quantity:
5 g
100 g
1 kg
361-09-1
C24H39NaO5
430.56
MFCD00150749,MFCD00064138
NRHMKIHPTBHXPF-TUJRSCDTSA-M
sodium cholate, sodium 3,7,12-trihydroxycholan-24-oate, ksc575s0b, 3,7,12-trihydroxycholan-24-oic acid sodium salt
57349315
sodium (4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate
[Na+].C[C@H](CCC([O-])=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C
361-09-1
C24H39NaO5
430.56
MFCD00150749,MFCD00064138
NRHMKIHPTBHXPF-TUJRSCDTSA-M
sodium cholate, sodium 3,7,12-trihydroxycholan-24-oate, ksc575s0b, 3,7,12-trihydroxycholan-24-oic acid sodium salt
57349315
sodium (4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate
[Na+].C[C@H](CCC([O-])=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C

Cholic Acid Sodium Salt, ≥98.5% is used in the solubilization of membrane-bound proteins in their native state.

  • Cholic Acid Sodium Salt, ≥98.5% is a component for the preparation of liposomes.
  • It has also been used in a study to investigate the elastic deformable vesicles, transfersomes, for transdermal drug delivery.
  • Critical Micelle Concentration: 14 mM
Color White
Physical Form Powder
Quantity 5 g
Formula Weight 430.6
Percent Purity ≥98%
Chemical Name or Material Cholic acid sodium salt
EINECSNumber 206-643-5
Recommended Storage Store at Room Temperature.

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit