Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Choline Acetyltransferase/ChAT Antibody - Azide and BSA Free, Novus Biologicals™
SDP

Sheep Polyclonal Antibody

$206.50

Specifications

Antigen Choline Acetyltransferase/ChAT
Concentration LYOPH
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB017939
SDP
View Documents
Novus Biologicals
NB100986930.1MG
0.1 mg
Each for $206.50
Only null left
Add to Cart
 
NB10098693
SDP
View Documents
Novus Biologicals
NB10098693
0.5 mg N/A N/A N/A
Description

Description

Choline Acetyltransferase/ChAT Polyclonal antibody specifically detects Choline Acetyltransferase/ChAT in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifications

Specifications

Choline Acetyltransferase/ChAT
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Western Blot
Unconjugated
Sheep
Q03059, P32738
1103
IgG
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
LYOPH
Polyclonal
Purified
RUO
acetyl CoA:choline O-acetyltransferase, ChAT, CHOACTase, Choline acetylase, choline acetyltransferase, choline O-acetyltransferase, CMS1A, CMS1A2, EC 2.3.1, EC 2.3.1.6
CHAT
Primary
ChAT.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.