Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Choline bitartrate, 97%, Acros Organics

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
87-67-2
C5H14NO·C4H5O6
253.25
QWJSAWXRUVVRLH-UHFFFAOYSA-M
choline bitartrate,cholini bitartras,choline 3-carboxy-2,3-dihydroxypropanoate,2-hydroxyethyl trimethylammonium bitartrate,choline tartrate 1:1,2-hydroxy-n,n,n-trimethylethanaminium 3-carboxy-2,3-dihydroxypropanoate,cholinibitatratis,choline hydrogen l-+-tartrate,ethanaminium, 2-hydroxy-n,n,n-trimethyl-, 2r,3r-2,3-dihydroxybutanedioate 1:1
10198924
2-hydroxyethyl(trimethyl)azanium;2,3,4-trihydroxy-4-oxobutanoate
C[N+](C)(C)CCO.C(C(C(=O)[O-])O)(C(=O)O)O
87-67-2
C5H14NO·C4H5O6
253.25
QWJSAWXRUVVRLH-UHFFFAOYSA-M
choline bitartrate,cholini bitartras,choline 3-carboxy-2,3-dihydroxypropanoate,2-hydroxyethyl trimethylammonium bitartrate,choline tartrate 1:1,2-hydroxy-n,n,n-trimethylethanaminium 3-carboxy-2,3-dihydroxypropanoate,cholinibitatratis,choline hydrogen l-+-tartrate,ethanaminium, 2-hydroxy-n,n,n-trimethyl-, 2r,3r-2,3-dihydroxybutanedioate 1:1
10198924
2-hydroxyethyl(trimethyl)azanium;2,3,4-trihydroxy-4-oxobutanoate
C[N+](C)(C)CCO.C(C(C(=O)[O-])O)(C(=O)O)O
Packaging Glass bottle
Melting Point 151.0°C to 153.0°C
Quantity 500g
Assay Percent Range 97%
Merck Index 15,2211
Solubility Information Solubility in water: soluble.
Formula Weight 253.25
Percent Purity 97%
Chemical Name or Material Choline bitartrate

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.

GHS Signal Word: Warning

EINECSNumber : 201-763-4