Learn More
Choline Bitartrate, FCC, 98%, Spectrum™
Click to view available options
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
C1228, 87-67-2, C9H19NO7
Spectrum™ Choline Bitartrate, FCC is an essential nutrient and typically grouped with B-complex vitamins. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Ignition Residue 0.001
Lead (Pb) 2 mg/kg
Packaging Poly Pail
Water 0.005