Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Choline hydroxide, 46% w/w aq. soln.

Manufacturer:  Alfa Aesar™ B24443AC

Quantity 25mL

 View more versions of this product

Catalog No. AAB24443AC

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

Applications
Choline hydroxide is used as a precursor molecule for the neurotransmitter acetylcholine, it is involved in several functions including muscle and memory muscle control. It was used as a basic catalyst for aldol condensation reactions.

Solubility
Fully miscible in water.

Notes
Store in a cool, dry place in tightly closed container. Store away from oxidizing agents, air, acids.
Description & Specifications

Chemical Identifiers

123-41-1
HOCH2CH2N(CH3)3OH
MFCD00002831
(2-Hydroxyethyl)trimethylammonium hydroxide
C5H15NO2
121.18
UN3267
Choline hydroxide

Specifications

123-41-1
HOCH2CH2N(CH3)3OH
MFCD00002831
(2-Hydroxyethyl)trimethylammonium hydroxide
25mL
C5H15NO2
121.18
UN3267
46% w/w aq. soln.
Choline hydroxide

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314
Causes severe skin burns and eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.

RTECSNumber : GA4025500

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit