Learn More
Thermo Scientific Chemicals Choline hydroxide, 45 wt.% aqueous solution, pure, stabilized
Supplier: Thermo Scientific Chemicals 386725000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Choline hydroxide, 45 wt.% aqueous solution | |
White | |
Glass Bottle | |
500 mL | |
15,2211 | |
choline hydroxide, gossypine, sincaline, bursine, luridine, sinkalin, sinkaline, fagine, vidine, 2-hydroxy-n,n,n-trimethylethanaminium hydroxide | |
KIZQNNOULOCVDM-UHFFFAOYSA-M | |
2-hydroxyethyl(trimethyl)azanium;hydroxide | |
121.18 | |
121.18 | |
Pure |
123-41-1, 7732-18-5 | |
1.0700g/mL | |
C5H15NO2 | |
MFCD00002831 | |
1.07 | |
Solubility in water: miscible. | |
[OH-].C[N+](C)(C)CCO | |
1800ppm ethylenediamine 3000ppm hydroxylamine | |
31255 | |
44.0 to 48.0% (Total base) | |
Granular Powder |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 204-625-1
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
RUO – Research Use Only