Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Choline Transporter Mouse anti-Mouse, Clone: 62-2E8, MilliporeSigma™

Mouse Monoclonal Antibody

$373.00

Specifications

Antigen Choline Transporter
Clone 62-2E8
Concentration 0.5 mg/mL
Applications Immunohistochemistry (Paraffin), ELISA
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB5514I100
View Documents
MilliporeSigma
MAB5514I100UL
100 μL
Each for $373.00
Add to cart
 
MAB5514I25
View Documents
MilliporeSigma
MAB5514I25UL
25 μL
Each for $373.00
Add to cart
 
Description

Description

Choline Transporter Monoclonal antibody specifically detects Choline Transporter in Mouse samples. It is validated for Immunohistochemistry (Paraffin), ELISA.
Specifications

Specifications

Choline Transporter
0.5 mg/mL
Unconjugated
Mouse
Mouse
GST-tagged recombinant fragment corresponding to 80 amino acids from the C-terminal region of human choline transporter.
Monoclonal
Protein G purified
62-2E8
Immunohistochemistry (Paraffin), ELISA
Purified
RUO
Purified monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
IgG1
Primary
Stable for 1 year at +2°C to +8°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit