Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chondroitin Sulfate Sodium Salt Dietary Supplement, USP, Spectrum™

Click to view available options
Quantity:
100 g
500 g
2.5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7/9/9082
7/9/9082

C1610, 9082-07-9

Spectrum™ Chondroitin Sulfate Sodium Salt, USP Dietary Supplement is the sodium salt of a sulfated glycosaminoglycan (GAG) composed of a chain of alternating sugars (N-acetylgalactosamine and glucuronic acid) and is used for osteoarthritis. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Chloride 0.5%
Ignition Residue 20%-30%
Loss on Drying 12%
Packaging Poly Bottle
Specific Rotation -20.0° to -30.0°
Quantity 100 g