Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chondroitin sulfate synthase 3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | Chondroitin sulfate synthase 3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP159673
|
Novus Biologicals
NBP159673 |
100 μL |
Each of 1 for $436.00
|
|
Description
Chondroitin sulfate synthase 3 Polyclonal specifically detects Chondroitin sulfate synthase 3 in Human samples. It is validated for Western Blot.Specifications
Chondroitin sulfate synthase 3 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
Carbohydrate synthase 2, Chondroitin glucuronyltransferase 3, chondroitin sulfate synthase 3, Chondroitin synthase 2, chondroitin synthase-2, ChSy-2, CHSY2, CSS3N-acetylgalactosaminyltransferase 3, EC 2.4.1.175, EC 2.4.1.226, Glucuronosyl-N-acetylgalactosaminyl-proteoglycan4-beta-N-acetylgalactosaminyltransferase II, N-acetylgalactosaminyl-proteoglycan 3-beta-glucuronosyltransferase 3 | |
CHSY3 | |
IgG | |
Affinity Purified | |
100 kDa |
Western Blot | |
Unconjugated | |
RUO | |
Q70JA7 | |
337876 | |
Synthetic peptides corresponding to CHSY-2 The peptide sequence was selected from the N terminal of CHSY-2. Peptide sequence PPLQQRRRGREPEGATGLPGAPAAEGEPEEEDGGAAGQRRDGRPGSSHNG. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title