Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Chondrolectin Monoclonal Antibody (396305)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA524131
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
This gene encodes a type I membrane protein with a carbohydrate recognition domain characteristic of C-type lectins in its extracellular portion. In other proteins, this domain is involved in endocytosis of glycoproteins and exogenous sugar-bearing pathogens. This protein localizes predominantly to the perinuclear region. Several transcript variants encoding a few different isoforms have been found for this gene.
Specifications
Chondrolectin | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9H9P2 | |
CHODL | |
Mouse myeloma cell line NS0-derived recombinant human Chondrolectin Arg22-Asn216. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry, Western Blot, Immunocytochemistry | |
396305 | |
Unconjugated | |
CHODL | |
3110074E07Rik; C21orf68; CHODL; Chondrolectin; FLJ12627; LOW QUALITY PROTEIN: chondrolectin; MT75; PRED12; Transmembrane protein MT75; UNQ872/PRO1890 | |
Mouse | |
Protein A/G | |
RUO | |
140578 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction