Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Chondrolectin Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109534
Description
Immunogen sequence: YRNWYTDEPS CGSEKCVVMY HQPTANPGLG GPYLYQWNDD RCNMKHNYIC KYEPEINPTA PVEKPYLTNQ PGDTHQNVVV TEAGIIPNLI YVVIPTIPLL L.
This gene encodes a type I membrane protein with a carbohydrate recognition domain characteristic of C-type lectins in its extracellular portion. In other proteins, this domain is involved in endocytosis of glycoproteins and exogenous sugar-bearing pathogens. This protein localizes predominantly to the perinuclear region. Several transcript variants encoding a few different isoforms have been found for this gene.
Specifications
Chondrolectin | |
Polyclonal | |
Unconjugated | |
CHODL | |
3110074E07Rik; C21orf68; CHODL; Chondrolectin; FLJ12627; LOW QUALITY PROTEIN: chondrolectin; MT75; PRED12; Transmembrane protein MT75; UNQ872/PRO1890 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
246048, 288289 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.25 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q9CXM0 | |
CHODL | |
Recombinant fusion protein containing a sequence corresponding to amino acids 110-210 of human CHODL (NP_001191103.1). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction