Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chordin-like 2/CHRDL2 Antibody (272905), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB24481MFV610
Description
Chordin-like 2/CHRDL2 Monoclonal antibody specifically detects Chordin-like 2/CHRDL2 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
| Chordin-like 2/CHRDL2 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Flow Cytometry, CyTOF | |
| 272905 | |
| Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| chordin-like 2, chordin-like protein 2 | |
| Mouse myeloma cell line NS0-derived recombinant human Chordin-like 2/CHRDL2, Arg28-Thr451, Accession # NP_056239 | |
| RUO | |
| 25884 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction