Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chordin Rat anti-Mouse, Alexa Fluor™ 750, Clone: RM0193-16B6, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212191AF750

Catalog No. NP212191A70


Add to cart

Description

Description

Chordin Monoclonal antibody specifically detects Chordin in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

Chordin
RM0193-16B6
Western Blot
CHRD, MGC133038
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse
Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
CHRD
Mouse Chordin
0.1 mL
Primary
8646
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit