Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chorionic Gonadotropin alpha Chain (alpha HCG) Mouse anti-Human, Clone: LHa/756- Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

$378.35

Specifications

Antigen Chorionic Gonadotropin alpha Chain (alpha HCG)
Clone LHa/756
Host Species Mouse
Isotype IgG1 Kappa
Gene Alias CGA glycoprotein hormones, alpha polypeptide, CG-ALPHA, FSHA, GPHa, HCG, LHA, TSHA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB254500100 View Documents Novus Biologicals
NBP254500100UG
100 ug Each for $378.35
Add to cart
 
NB254500200 View Documents Novus Biologicals
NBP254500200UG
200 ug This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Chorionic Gonadotropin alpha Chain (hCG alpha) Monoclonal antibody specifically detects Chorionic Gonadotropin alpha Chain (hCG alpha) in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications

Specifications

Chorionic Gonadotropin alpha Chain (alpha HCG)
Mouse
CGA glycoprotein hormones, alpha polypeptide, CG-ALPHA, FSHA, GPHa, HCG, LHA, TSHA
Immunohistochemistry (Paraffin)
Unconjugated
Store at -20 to -80C. Avoid freeze-thaw cycles.
Monoclonal
1 mg/mL
LHa/756
IgG1 Kappa
Human
RUO
Purified
Primary
PBS with No Preservative

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit